Learn More
Michigan Residual Solvents Mixture 470 100-1000 μg/mL in Triacetin, Dr. Ehrenstorfer
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
Specifications
Specifications
| Chemical Name or Material | 1,2-Dichloroethane∣2,2-Dimethylbutane∣2,3-Dimethylbutane∣2-Methylbutane∣2-Methylpentane∣3-Methylpentane∣Acetone∣Acetonitrile∣Benzene∣Chloroform∣Dichloromethane (Methylenechloride)∣Diethylether∣Ethanol∣Ethyl acetate∣Isopropyl alcohol∣Methanol∣n-Heptane∣n-Hexane∣n-Pentane∣Toluene∣Trichloroethene∣Xylene (all isomers) |
| CAS | 107-06-2, 75-83-2, 79-29-8, 78-78-4, 107-83-5, 96-14-0 |
| Analyte or Component Names | 1,2-Dichloroethane∣2,2-Dimethylbutane∣2,3-Dimethylbutane∣2-Methylbutane∣2-Methylpentane∣3-Methylpentane∣Acetone∣Acetonitrile∣Benzene∣Chloroform∣Dichloromethane (Methylenechloride)∣Diethylether∣Ethanol∣Ethyl acetate∣Isopropyl alcohol∣Methanol∣n-Heptane∣n-Hexane∣n-Pentane∣Toluene∣Trichloroethene∣Xylene (all isomers) |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Triacetin |
| Industry Type | Food Testing |
| Quantity | 5 x 1 mL |
| Concentration or Composition (by Analyte or Components) | 1,2-Dichloroethane 100 μg/mL∣Benzene 100 μg/mL∣Chloroform 100 μg/mL∣Trichloroethene 100 μg/mL∣2,2-Dimethylbutane 1000 μg/mL∣2,3-Dimethylbutane 1000 μg/mL∣2-Methylbutane 1000 μg/mL∣2-Methylpentane 1000 μg/mL∣3-Methylpentane 1000 μg/mL∣Acetone 1000 μg/mL∣Acetonitrile 1000 μg/mL∣Dichloromethane (Methylenechloride) 1000 μg/mL∣Diethylether 1000 μg/mL∣Ethanol 1000 μg/mL∣Ethyl acetate 1000 μg/mL∣Isopropyl alcohol 1000 μg/mL∣Methanol 1000 μg/mL∣n-Heptane 1000 μg/mL∣n-Hexane 1000 μg/mL∣n-Pentane 1000 μg/mL∣Toluene 1000 μg/mL∣Xylene (all isomers) 1000 μg/mL |