missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICALCL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00€ - 624.00€
Specifications
| Antigen | MICALCL |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18609225
|
Novus Biologicals
NBP2-38455-25ul |
25 μL |
280.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18112368
|
Novus Biologicals
NBP2-38455 |
0.1 mL |
624.00€
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MICALCL Polyclonal specifically detects MICALCL in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| MICALCL | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q6ZW33 | |
| 84953 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: KKLVLTQEQKTMLLDWNDSIPESVHLKAGERISQKSAENGRGGRVLKPVRPLLLPRAAGEPLPTQRGAQEKMGTPAEQAQGE | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ebitein-1, Ebitein1, ERK2-binding testicular protein 1, FLJ14966, MICAL C-terminal like, MICAL C-terminal-like protein | |
| MICALCL | |
| IgG | |
| Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title