Learn More
IBA Lifesciences MHC I-Strep HLA-A*0201; CMV IE-1 (VLEETSVML)
GREENER_CHOICE
Product Code. 15693918 Shop All IBA Lifesciences Products
Click to view available options
:
Capacity: 40μL
Product Code. 15693918
Brand: IBA Lifesciences 67041001
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
MHC class I molecule fused to a Twin-Strep-tag™ and refolded with a peptide
  • MHC I-Strep HLA-A*0201; CMV IE-1 (316-324) (VLEETSVML) is a recombinantly expressed MHC class I molecule that presents a peptide from a human cytomegalovirus (CMV) Immediate-Early (IE).
  • Our MHC I Streptamer™ approach is based on our Strep-tag technology and facilitates reversible antigen-specific T cell labeling.
  • A Twin-Strep-tag™ fused to the MHC I-Strep permits multimerization of MHC I complexes on a Strep-Tactin™ backbone, resulting in the formation of MHC I Streptamers.
  • For multimerizing MHC I-Streps, Strep-Tactin™ TACS Agarose columns, Strep-Tactin™ Magnetic Microbeads or fluorophore-conjugated Strep-Tactin™ can be used.
  • This allows affinity chromatographic or magnetic isolation or cell staining for flow cytometry analysis and sorting (FACS), depending on the experimental requirements.
  • The addition of biotin reverses the multimerization of MHC class I molecules, leading to their dissociation from the target cells.
  • The resulting label-free cells are highly viable and fully functional.
  • Specifications
    Capacity (Metric) 40 μL
    Kit Contents Fluorescent reagent for staining, magnetic T cell labeling and isolation