missing translation for 'onlineSavingsMsg'
Learn More

MHC Class II antigen I Ak,d,b,q,r Antibody (ER-TR3), DyLight 680, Novus Biologicals™

Product Code. 18028343 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.125 mL
Unit Size:
125µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18028343 0.125 mL 125µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18028343 Supplier Novus Biologicals Supplier No. NB10064961FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

MHC Class II antigen I Ak,d,b,q,r Monoclonal specifically detects MHC Class II antigen I Ak,d,b,q,r in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class II antigen I Ak,d,b,q,r
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone ER-TR3
Conjugate DyLight 680
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Gene Alias Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly
Gene Symbols HLA-DRB1
Host Species Rat
Immunogen Isolated C3H thymic stromal cells
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3123
Test Specificity NB100-64961 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class II molecule. In the mouse, MHC class II molecules are expressed by dendritic cells, B-cells and macrophages. In thymus tissues, clone ER-TR3 stains cortical and medullary stromal cells. Clone ER-TR3 specifically recognizes mouse strains with the haplotypes I-A k,b,d,q,r. Mouse strains with the haplotypes I-A s,f are not recognized by this antibody.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.