missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II antigen I Ak,d,b,q,r Antibody (ER-TR3), DyLight 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64961DL594
481.95 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
MHC Class II antigen I Ak,d,b,q,r Monoclonal antibody specifically detects MHC Class II antigen I Ak,d,b,q,r in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| MHC Class II antigen I Ak,d,b,q,r | |
| Monoclonal | |
| DyLight 594 | |
| Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly | |
| Isolated C3H thymic stromal cells | |
| 0.125 mL | |
| Immunology | |
| 3123 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| ER-TR3 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction