missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II antigen I Ak,d,b,q,r Antibody (ER-TR3), Allophycocyanin, Novus Biologicals™
Shop All Bio Techne ProductsDescription
MHC Class II antigen I Ak,d,b,q,r Monoclonal specifically detects MHC Class II antigen I Ak,d,b,q,r in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
| Antigen | MHC Class II antigen I Ak,d,b,q,r |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | ER-TR3 |
| Conjugate | APC |
| Dilution | Flow Cytometry |
| Gene Alias | Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly |
| Gene Symbols | HLA-DRB1 |
| Host Species | Rat |
| Immunogen | Isolated C3H thymic stromal cells |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?