missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Antibody (ER-TR2), DyLight 650, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NB100-64959C
488.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
MHC Class II Monoclonal antibody specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| MHC Class II | |
| Monoclonal | |
| DyLight 650 | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| Isolated C3H thymic stromal cells | |
| 0.125 mL | |
| Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators | |
| 3108 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| ER-TR2 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction