missing translation for 'onlineSavingsMsg'
Learn More

MHC Class I Antibody (R1-21.2) - Chimeric - Azide and BSA Free, Novus Biologicals™

Product Code. 18667116 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
Unit Size:
0.2mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18667116 0.2 mg 0.2mg
18087725 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18667116 Supplier Novus Biologicals Supplier No. NBP3090170.2mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MHC Class I Monoclonal antibody specifically detects MHC Class I in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmune Assays
TRUSTED_SUSTAINABILITY

Specifications

Antigen MHC Class I
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Radioimmune Assays (RIA)
Classification Monoclonal
Clone R1-21.2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Radioimmunoassay
Formulation PBS
Gene Alias DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Host Species Rabbit
Immunogen This antibody was raised by immunising Lewis 1F rats with mouse CH-1 lymphoma cells. Spleen cells from immunised rats were fused with X-63Ag8-653 myeloma cells to generate hybridomas.
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3133
Target Species Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.