Learn More
Abnova™ MGST1 Recombinant Protein
Description
The MAPEG (Membrane Associated Proteins in Eicosanoid and Glutathione metabolism) family consists of six human proteins, two of which are involved in the production of leukotrienes and prostaglandin E, important mediators of inflammation. Other family members, demonstrating glutathione S-transferase and peroxidase activities, are involved in cellular defense against toxic, carcinogenic, and pharmacologically active electrophilic compounds. This gene encodes a protein that catalyzes the conjugation of glutathione to electrophiles and the reduction of lipid hydroperoxides. This protein is localized to the endoplasmic reticulum and outer mitochondrial membrane where it is thought to protect these membranes from oxidative stress. Four transcript variants of this gene encode one protein isoform.
- Theoretical MW: 42.79kDa
- Preparation Method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Molecular Weight (g/mol) | 42.8 |
| Name | Human MGST1 Full-length ORF Recombinant Protein with GST-tag at N-terminal |
| pH Range | 8 |
| Preparation Method | In vitro wheat germ expression system |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE stained with Coomassie Blue |
| Quantity | 25 μg |
| Source | Wheat Germ (in vitro) |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.