missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MGME1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
183.00€ - 322.00€
Specifications
| Antigen | MGME1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632178
|
Novus Biologicals
NBP3-12644-50ul |
50 μg |
183.00€
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677717
|
Novus Biologicals
NBP3-12644-100ul |
100 μg |
322.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MGME1 Polyclonal antibody specifically detects MGME1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| MGME1 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human | |
| bA504H3.4, chromosome 20 open reading frame 72, FLJ14597, hypothetical protein LOC92667 | |
| Produced in rabbits immunized with E. coli-derived Human MGME1 fragment | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5ul/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, pH7.0 | |
| 92667 | |
| IgG | |
| Antigen and protein A Affinity-purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title