missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NLS921
This item is not returnable.
View return policy
Description
mGluR7 Polyclonal antibody specifically detects mGluR7 in Human, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Specifications
| mGluR7 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1.0 mg/mL | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 6 - 16 μg/mL, Immunocytochemistry 1:10 - 1:500 | |
| FLJ40498, glutamate receptor, metabotropic 7, metabotropic glutamate receptor 7, mGlu7, mGluR7, MGLUR7GPRC1GGLUR7 | |
| Synthetic 19 amino acid peptide from N-terminal extracellular domain of human mGluR7. | |
| 0.05 mL | |
| GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
| 2917 | |
| Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction