missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mGluR5/1a Antibody (S75-33), Novus Biologicals™
Mouse Monoclonal Antibody
166.00€ - 543.00€
Specifications
| Antigen | mGluR5/1a |
|---|---|
| Clone | S75-33 |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18235285
|
Novus Biologicals
NBP2-59334 |
100 μg |
543.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646166
|
Novus Biologicals
NBP2-59334-25ug |
25 μg |
166.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mGluR5/1a Monoclonal specifically detects mGluR5/1a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| mGluR5/1a | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b | |
| Primary | |
| Detects 130kDa (both mGluR1 and mGluR5). |
| S75-33 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title