Learn More
mGluR3 Antibody (SR1543), R&D Systems™
Product Code. 18611904 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18611904 50 μg 50µL
18633985 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18611904 Supplier Novus Biologicals Supplier No. NBP32238250ul
 Restricted Item
Estimated Shipment: 07-12-2026
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
mGluR3 Monoclonal specifically detects mGluR3 in Human∣Mouse∣Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Antigen mGluR3
Applications Western Blot
Classification Monoclonal
Clone SR1543
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias GLUR3, glutamate receptor, metabotropic 3, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3
Host Species Rabbit
Immunogen A synthesized peptide derived from human mGluR3 (Uniprot #: Q14832)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2913
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus