missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ mGluR2 Monoclonal Antibody (421614)
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Glutamate is the main excitatory neurotransmitter in the brain. For many years it had been considered to act only on the ligand-gated receptor channels-termed NMDA, AMPA and kainite receptors that are involved in the fast excitatory synaptic transmission. Recently, glutamate has been shown to regulate enzymes producing second messengers via specific receptors coupled to G-proteins. These receptors are called metabotropic glutamate receptors. In expression systems, Group-I receptors stimulate phospholipase C as revealed by an increase in phosphoinositide turnover and calcium release from internal stores. Group-II and -III receptors are coupled to the inhibition of adenylyl cyclase. The Group-I receptors include mGluR1a and mGluR5. The Group-II receptors in include mGluR2 and mGluR3.
Specifications
Specifications
| Antigen | mGluR2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | 421614 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and no preservative |
| Gene | GRM2 |
| Gene Accession No. | Q14416 |
| Gene Alias | 4930441L02Rik; G protein coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member B; GLUR2; glutamate metabotropic receptor 2; glutamate receptor; glutamate receptor homolog; glutamate receptor, metabotropic 2; GPRC1B; Grm2; M mGlu2; Metabotropic; Metabotropic glutamate receptor 2; mGlu2; MGLUR2; mGluR7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?