missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MFRP Monoclonal Antibody (291218)

Rat Monoclonal Antibody

Brand:  Invitrogen™ MA561062

Product Code. 30533710

  • 429.00€ / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

MFRP is a member of the frizzled-related proteins. It may play a role in eye development, as mutations in this gene have been associated with nanophthalmos, posterior microphthalmia, retinitis pigmentosa, foveoschisis, and optic disc drusen.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MFRP
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
Q9BY79
MFRP
Mouse myeloma cell line NS0-derived recombinant human MFRP, Ser101-Pro579.
100 μg
Primary
Human
Antibody
IgG1
Western Blot
291218
Unconjugated
MFRP
C1QTNF5; DKFZp586B0621; FLJ30570; MCOP5; Membrane frizzled-related protein; membrane-type frizzled-related protein; MFRP; MGC32938; NNO2; rd6
Rat
Protein A/G
RUO
83552
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.