missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ MFI2 Recombinant Protein

Product Code. 16157781
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy

Product Code. 16157781

Brand: Abnova™ H00004241P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant protein for gene encoding cell-surface glycoprotein found on melanoma cells

The protein shares sequence similarity and iron-binding properties with members of the transferrin superfamily. The importance of the iron binding function has not yet been identified. This gene resides in the same region of chromosome 3 as members of the transferrin superfamily. Alternative splicing results in two transcript variants.

  • Human MFI2 full-length ORF ( AAH01875, 21 a.a. - 302 a.a.) recombinant protein with GST-tag at N-terminal
  • Gene description: antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and 96.5
  • Theoretical molecular weight: 56.76kDa
  • Preparation method: in vitro wheat germ expression system
  • Purification: glutathione sepharose 4 fast flow
  • Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer

Sequence: MEVRWCATSDPEQHKCGNMSEAFREAGIQPSLLCVRGTSADHCVQLIAAQEADAITLDGGAIYEAGKEHGLKPVVGEVYDQEVGTSYYAVAVVRRSSHVTIDTLKGVKSCHTGINRTVGWNVPVGYLVESGRLSVMGCDVLKAVSDHFGGSCVPGAGETSYSESLCRLCRGDSPGEGVCDK SPLERYYDYSGAFRCLAEGAGDVAFVKHSTVLENTDESPSRRQTWTRSEEEEGECPAHEEARRTMRSSAGQAWKWAPVHRPQDESDKGEFGKRAKSRDMLG

Best use within three months from the date of receipt.

ELISA, western blotting (recombinant protein), antibody production, protein array

Specifications

Accession Number AAH01875
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Gene ID (Entrez) 4241
Molecular Weight (g/mol) 56.76
Name MFI2 (Human) Recombinant Protein (P01)
pH Range 8
Preparation Method In vitro wheat germ expression system
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Source Wheat Germ (in vitro)
Immunogen MEVRWCATSDPEQHKCGNMSEAFREAGIQPSLLCVRGTSADHCVQLIAAQEADAITLDGGAIYEAGKEHGLKPVVGEVYDQEVGTSYYAVAVVRRSSHVTIDTLKGVKSCHTGINRTVGWNVPVGYLVESGRLSVMGCDVLKAVSDHFGGSCVPGAGETSYSESLCRLCRGDSPGEGVCDKSPLERYYDYSGAFRCLAEGAGDVAFVKHSTVLENTDESPSRRQTWTRSEEEEGECPAHEEARRTMRSSAGQAWKWAPVHRPQDESDKGEFGKRAKSRDMLG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CD228/FLJ38863/MAP97/MGC4856/MTF1
Common Name MFI2
Gene Symbol MFI2
Cross Reactivity Human
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Form Solution
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.