Learn More
METTL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15073-1-AP-150UL
This item is not returnable.
View return policy
Description
METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.Specifications
| METTL3 | |
| Polyclonal | |
| Unconjugated | |
| METTL3 | |
| IME4, M6A, methyltransferase like 3, METTL3, MT A70, MTA70, Spo8 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 361035, 56335, 56339 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q86U44, Q8C3P7 | |
| Mettl3 | |
| METTL3 Fusion Protein Ag7110 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |