missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ METRNL Monoclonal Antibody (829535)

Rat Monoclonal Antibody
Marca: Invitrogen™ MA561705
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute at 0.5 mg/mL in sterile PBS.
Hormone induced following exercise or cold exposure that promotes energy expenditure. Induced either in the skeletal muscle after exercise or in adipose tissue following cold exposure and is present in the circulation. Able to stimulate energy expenditure associated with the browning of the white fat depots and improves glucose tolerance. Does not promote an increase in a thermogenic gene program via direct action on adipocytes, but acts by stimulating several immune cell subtypes to enter the adipose tissue and activate their prothermogenic actions. Stimulates an eosinophil-dependent increase in IL4 expression and promotes alternative activation of adipose tissue macrophages, which are required for the increased expression of the thermogenic and anti-inflammatory gene programs in fat. Required for some cold-induced thermogenic responses, suggesting a role in metabolic adaptations to cold temperatures.
Especificaciones
| METRNL | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q8VE43 | |
| METRNL | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse METRNL, Gln46-Glu311. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Flow Cytometry | |
| 829535 | |
| Unconjugated | |
| METRNL | |
| 9430048M07Rik; BC019776; cometin; meteorin like, glial cell differentiation regulator; meteorin, glial cell differentiation regulator-like; meteorin-like protein; METRNL; Subfatin | |
| Rat | |
| Protein A/G | |
| RUO | |
| 210029 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido