missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metoprolol Fumarate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Metoprolol Fumarate
Specifications
Specifications
| Chemical Name or Material | Metoprolol Fumarate |
| CAS | 80274-67-5 |
| Analyte or Component Names | Metoprolol Fumarate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C15 H25 N O3 . C4 H4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol (2E)-2-butenedioate (2:1) (salt), CGP 2175C, Lopressor OROS, Metoprolol fumarate, (+/-)-1-[4-(2-Methoxyethyl)phenoxy]-3-[(1-methylethyl)amino]-2-propanol (E)-2-butenedioate (2:1) (salt) |
| InChI Formula | InChI=1S/2C15H25NO3.C4H4O4/c2*1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;5-3(6)1-2-4(7)8/h2*4-7,12,14,16-17H,8-11H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+ |
| SMILES | COCCc1ccc(OCC(O)CNC(C)C)cc1.COCCc2ccc(OCC(O)CNC(C)C)cc2.OC(=O)\C=C\C(=O)O |
| IUPAC Name | (E)-but-2-enedioic acid;1-[4-(2-methoxyethyl)phenoxy]-3-(propan-2-ylamino)propan-2-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction