Learn More
Thermo Scientific Chemicals Methyl Red
Product Code. p-7023292
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. 11422688
Brand: Thermo Scientific Chemicals A16690.22
Request Bulk or Custom Format
CAS: 493-52-7 | C15H15N3O2 | 269.304 g/mol
Bacterial strains isolated from dye-contaminated sludge decolorized a toxic azo dye, namely, methyl red (MR). Methyl red is used in the methyl red test (MR test), used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bacterial strains isolated from dye-contaminated sludge decolorized a toxic azo dye, namely, methyl red (MR). Methyl red is used in the methyl red test (MR test), used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose
Solubility
Insoluble in water. Soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 493-52-7
Molecular Formula C15H15N3O2
Molecular Weight (g/mol) 269.304
MDL Number MFCD00002425
InChI Key CEQFOVLGLXCDCX-UHFFFAOYSA-N
Synonym C.I. 13020
PubChem CID 10303
IUPAC Name 2-[[4-(dimethylamino)phenyl]diazenyl]benzoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)O
Specifications
Melting Point 179°C to 182°C
Beilstein 750102
Merck Index 14,6119
Solubility Information Insoluble in water. Soluble in ethanol.
Formula Weight 269.3
Quantity 100 g
Chemical Name or Material Methyl Red