Learn More
Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate, 95%, Thermo Scientific Chemicals
Product Code. 10525134 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10525134 1 g Glass bottle 1g
10625125 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10525134 Supplier Thermo Scientific Acros Supplier No. 426350010
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 95715-86-9
Molecular Formula C12H21NO5
Molecular Weight (g/mol) 259.30
MDL Number MFCD00674041
InChI Key ZNBUXTFASGDVCL-UHFFFAOYNA-N
Synonym r-3-tert-butyl 4-methyl 2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylate, 3-tert-butyl 4-methyl 4r-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate, r-+-3-tert-butoxycarbonyl-2,2-dimethyl-4-oxazolidinecarboxylic acid methyl ester, methyl r-n-boc-2,2-dimethyloxazolidine-4-carboxylate, r-+-3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, r---3-boc-4-methoxycarbonyl-2,2-dimethyl-1,3-oxazolidine, methyl r-+-3-boc-2,2-dimethyl-4-oxazolidinecarboxylate, 3-tert-butyl 4-methyl r-+-2,2-dimethyloxazolidine-3,4-dicarboxylate, methyl 3-tert-butoxycarbonyl-2,2-dimethyloxazolidine-4-r-carboxylate
PubChem CID 688220
IUPAC Name 3-O-tert-butyl 4-O-methyl (4R)-2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
SMILES COC(=O)C1COC(C)(C)N1C(=O)OC(C)(C)C
Specifications
Density 1.0820g/mL
Boiling Point 89.0°C (0.8 mmHg)
Flash Point 93°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 1 g
Specific Gravity 1.082
Formula Weight 259.3
Percent Purity 95%
Chemical Name or Material Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate
compliance-icons
Product Identifier
  • Methyl (R)-(+)-3-BOC-2,2-dimethyl-4-oxazolidinecarboxylate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.