accessibility menu, dialog, popup

UserName

CAS RN 95715-86-9

H3CNOOOCH3OOCH3CH3H3CCH3

CAS RN 95715-86-9

IUPAC Name: 3-tert-butyl 4-methyl 2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
Synonyms: 3-tert-butyl 4-methyl 2,2-dimethyl-1,3-oxazolidine-3,4-dicarboxylate
Molecular Weight (g/mol): 259.3
Molecular Formula: C12H21NO5
InChi Key: ZNBUXTFASGDVCL-UHFFFAOYNA-N
SMILES: COC(=O)C1COC(C)(C)N1C(=O)OC(C)(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results