missing translation for 'onlineSavingsMsg'
Learn More

Methyl Paraben-13C6, TRC

Product Code. 30012637
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012637 10 mg 10mg
30012037 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012637 Supplier Toronto Research Chemicals Supplier No. TRCM32566410MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1581694-95-2
Molecular Formula C213C6H8O3
Molecular Weight (g/mol) 158.1
Synonym 4-Hydroxybenzoic Acid Methyl Ester-13C6, 4-(Carbomethoxy)phenol-13C6, 4-(Methoxycarbonyl)phenol-13C6, 4-Hydroxybenzoic Acid Methyl Ester-13C6, 4-Hydroxymethyl Benzoate-13C6, Danisol M-13C6, E 218-13C6, Killitol-13C6, Maseptol-13C6, Mekkings M-13C6, Metaben-13C6, Metagin-13C6, Paridol-13C6, Preserval-13C6, Solbrol-13C6;
SMILES O[13C]1=[13CH][13CH]=[13C](C(OC)=O)[13CH]=[13CH]1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H8O3/c1-11-8(10)6-2-4-7(9)5-3-6/h2-5,9H,1H3/i2+1,3+1,4+1,5+1,6+1,7+1
Percent Purity >95%
Chemical Name or Material Methyl Paraben-13C6
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.