CAS RN 1581694-95-2
CAS RN 1581694-95-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Methyl Paraben-13C6, TRC
CAS: 1581694-95-2 Molecular Formula: C213C6H8O3 Molecular Weight (g/mol): 158.1 Synonym: 4-Hydroxybenzoic Acid Methyl Ester-13C6,4-(Carbomethoxy)phenol-13C6,4-(Methoxycarbonyl)phenol-13C6,4-Hydroxybenzoic Acid Methyl Ester-13C6,4-Hydroxymethyl Benzoate-13C6,Danisol M-13C6,E 218-13C6,Killitol-13C6,Maseptol-13C6,Mekkings M-13C6,Metaben-13C6,Metagin-13C6,Paridol-13C6,Preserval-13C6,Solbrol-13C6; SMILES: O[13C]1=[13CH][13CH]=[13C](C(OC)=O)[13CH]=[13CH]1
2
Methyl 4-hydroxybenzoate-ring-13C6 solution, 10 μg/mL in acetone, Honeywell Fluka™
10 μg/mL in acetone. Analytical standard, reference material