Learn More
Methyl L-xylo-Hex-2-ulosonate (Methyl L-Sorbosonate), Mikromol™
Description
Methyl L-xylo-Hex-2-ulosonate (Methyl L-Sorbosonate)
Specifications
Specifications
| Chemical Name or Material | Methyl L-Sorbosonate |
| CAS | 3031-98-9 |
| Analyte or Component Names | Methyl L-Sorbosonate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H12 O7 |
| Certifications/Compliance | ISO 9001 |
| Synonym | L-xylo-2-Hexulosonic acid, methyl ester, Gulonic acid, 2-keto-, methyl ester, L- (6CI), L-xylo-Hexulosonic acid, methyl ester (7CI,8CI), 2-keto-L-Gulonic acid methyl ester, Methyl 2-keto-L-gulonate, Methyl L-xylo-hexulosonate, NSC 163924, Methyl L-xylo-Hex-2-ulosonate, Ascorbic Acid Imp. D (EP), Methyl L-xylo-Hex-2-ulosonate, Ascorbic Acid Impurity D, Sodium Ascorbate Impurity D |
| InChI Formula | InChI=1S/C7H12O7/c1-14-7(13)6(12)5(11)4(10)3(9)2-8/h3-5,8-11H,2H2,1H3/t3-,4+,5-/m0/s1 |
| SMILES | COC(=O)C(=O)[C@@H](O)[C@H](O)[C@@H](O)CO |
| IUPAC Name | methyl (3S,4R,5S)-3,4,5,6-tetrahydroxy-2-oxo-hexanoate |
| Show More |