Learn More
Methyl [5-Propylsulphinyl)-1H-benzimidazol-2-yl]carbamate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Methyl [5-Propylsulphinyl)-1H-benzimidazol-2-yl]carbamate
Specifications
Specifications
| Chemical Name or Material | Albendazole sulfoxide |
| CAS | 54029-12-8 |
| Analyte or Component Names | Albendazole sulfoxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H15 N3 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Carbamic acid, [5-(propylsulfinyl)-1H-benzimidazol-2-yl]-, methyl ester (9CI), (±)-Albendazole sulfoxide, Albendazole oxide, Methyl [5-(propylsulfinyl)-1H-benzimidazol-2-yl]carbamate, RS 8852, Ricobendazole, Rycobendazole, Albendazole Imp. B (EP), SKF-77664, Ph Eur Albendazole Impurity B, Albendazole Impurity B |
| InChI Formula | InChI=1S/C12H15N3O3S/c1-3-6-19(17)8-4-5-9-10(7-8)14-11(13-9)15-12(16)18-2/h4-5,7H,3,6H2,1-2H3,(H2,13,14,15,16) |
| SMILES | CCCS(=O)c1ccc2[nH]c(NC(=O)OC)nc2c1 |
| IUPAC Name | methyl N-(5-propylsulfinyl-1H-benzimidazol-2-yl)carbamate |
| Show More |