missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-bromovalerate, 97%

Product Code. 11429376
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11429376

Brand: Thermo Scientific Alfa Aesar A19072.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5454-83-1 | C6H11BrO2 | 195.06 g/mol

It is employed as an intermediate in the production of Apixaban .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an intermediate in the production of Apixaban .

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5454-83-1
Molecular Formula C6H11BrO2
Molecular Weight (g/mol) 195.06
MDL Number MFCD00000265
InChI Key RAVVJKCSZXAIQP-UHFFFAOYSA-N
PubChem CID 79557
IUPAC Name methyl 5-bromopentanoate
SMILES COC(=O)CCCCBr

Specifications

Density 1.363
Boiling Point 79°C to 80°C (4 mmHg)
Flash Point 99°C (210°F)
Refractive Index 1.463
Quantity 25 g
Beilstein 1749649
Synonym methyl 5-bromovalerate, 5-bromovaleric acid methyl ester, pentanoic acid, 5-bromo-, methyl ester, methyl .omega.-bromopentanoate, 5-bromopentanoic acid methyl ester, 5-bromo-pentanoic acid methyl ester, methyl omega-bromopentanoate, methyl5-bromovalerate, valeric acid, 5-bromo-, methyl ester, methyl-5-bromovalerate
Solubility Information Slightly soluble in water.
Formula Weight 195.06
Percent Purity 97%
Chemical Name or Material Methyl 5-bromovalerate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.