CAS RN 5454-83-1
CAS RN 5454-83-1
IUPAC Name:
methyl 5-bromopentanoate
Synonyms:
methyl 5-bromopentanoate
Molecular Weight (g/mol):
195.06
Molecular Formula:
C6H11BrO2
InChi Key:
RAVVJKCSZXAIQP-UHFFFAOYSA-N
SMILES:
COC(=O)CCCCBr
1
–
2
of
2
results
1
Methyl 5-bromovalerate, 97%
CAS: 5454-83-1 Molecular Formula: C6H11BrO2 Molecular Weight (g/mol): 195.06 MDL Number: MFCD00000265 InChI Key: RAVVJKCSZXAIQP-UHFFFAOYSA-N PubChem CID: 79557 IUPAC Name: methyl 5-bromopentanoate SMILES: COC(=O)CCCCBr
2
Methyl 5-bromovalerate, 97%
CAS: 5454-83-1 Molecular Formula: C6H11BrO2 Molecular Weight (g/mol): 195.06 MDL Number: MFCD00000265 InChI Key: RAVVJKCSZXAIQP-UHFFFAOYSA-N PubChem CID: 79557 IUPAC Name: methyl 5-bromopentanoate SMILES: COC(=O)CCCCBr