missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-methylsulfonylpyrimidine-5-carboxylate, Mikromol™

Product Code. 30049675
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049675 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049675 Supplier Mikromol Supplier No. MM0936.360025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-methylsulfonylpyrimidine-5-carboxylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-(methylsulfonyl)pyrimidine-5-carboxylate
CAS 799842-06-1
Analyte or Component Names Methyl 4-(4-Fluorophenyl)-6-isopropyl-2-(methylsulfonyl)pyrimidine-5-carboxylate
Shipping Condition Room Temperature
Molecular Formula C16H17FN2O4S
Certifications/Compliance ISO 9001
Synonym 4-(4-Fluorophenyl)-6-(1-methylethyl)-2-(methylsulfonyl)-5-pyrimidinecarboxylic Acid Methyl Ester, 5-Pyrimidinecarboxylic acid, 4-(4-fluorophenyl)-6-(1-methylethyl)-2-(methylsulfonyl)-, methyl ester, Methyl 4-(4-fluorophenyl)-6-(1-methylethyl)-2-(methylsulfonyl)-5-pyrimidinecarboxylate, 4-(4-Fluorophenyl)-6-isopropyl-2-(methanesulfonyl)pyrimidine-5-carboxylic acid methyl ester
InChI Formula InChI=1S/C16H17FN2O4S/c1-9(2)13-12(15(20)23-3)14(10-5-7-11(17)8-6-10)19-16(18-13)24(4,21)22/h5-9H,1-4H3
SMILES COC(=O)c1c(nc(nc1c2ccc(F)cc2)S(=O)(=O)C)C(C)C
IUPAC Name methyl 4-(4-fluorophenyl)-2-methylsulfonyl-6-propan-2-ylpyrimidine-5-carboxylate
Molecular Weight (g/mol) 352.38
Quantity 25 mg
Formula Weight 352.0893 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.