Learn More
Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (Δ-1-Aza Ester), Mikromol™
Description
Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (Delta-1-Aza Ester), Mikromol
Specifications
Specifications
| Chemical Name or Material | Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (delta-1-Aza Ester) |
| CAS | 103335-41-7 |
| Analyte or Component Names | Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (delta-1-Aza Ester) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H29 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate, Finasteride Imp. B (EP), Delta-1-Aza Ester of Finasteride, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, methyl ester, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxylic acid, 3-oxo-, methyl ester, (5alpha,17beta)-, 17beta-(Methoxycarbonyl)-4-aza-5alpha-androst-1-en-3-one, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 1,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, methyl ester, [4aR-(4aalpha,4bbeta,6aalpha,7alpha,9abeta,9balpha,11abeta)]-, 3-Oxo-4-aza-5alpha-androst-1-en-17beta-carboxylic acid methyl ester, Methyl 3-oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate, Finasteride Impurity B |
| InChI Formula | InChI=1S/C20H29NO3/c1-19-10-8-14-12(13(19)5-6-15(19)18(23)24-3)4-7-16-20(14,2)11-9-17(22)21-16/h9,11-16H,4-8,10H2,1-3H3,(H,21,22)/t12-,13-,14-,15+,16+,19-,20+/m0/s1 |
| Solution Type | Neat |
| SMILES | COC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C |
| Show More |