missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (Δ-1-Aza Ester), Mikromol™

Product Code. 17964546
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17964546 100 mg 100mg
17961126 25 mg 25mg
30463330 4 x 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17964546 Supplier Mikromol Supplier No. MM0415.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (Δ-1-Aza Ester)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (Δ-1-Aza Ester)
CAS 103335-41-7
Analyte or Component Names Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate (Δ-1-Aza Ester)
Shipping Condition Room Temperature
Molecular Formula C20 H29 N O3
Certifications/Compliance ISO 17025
Synonym Methyl 3-Oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate, Finasteride Imp. B (EP), Delta-1-Aza Ester of Finasteride, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 2,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, methyl ester, (4aR,4bS,6aS,7S,9aS,9bS,11aR)-, 4-Azaandrost-1-ene-17-carboxylic acid, 3-oxo-, methyl ester, (5alpha,17beta)-, 17beta-(Methoxycarbonyl)-4-aza-5alpha-androst-1-en-3-one, 1H-Indeno[5,4-f]quinoline-7-carboxylic acid, 1,4a,4b,5,6,6a,7,8,9,9a,9b,10,11,11a-tetradecahydro-4a,6a-dimethyl-2-oxo-, methyl ester, [4aR-(4aalpha,4bbeta,6aalpha,7alpha,9abeta,9balpha,11abeta)]-, 3-Oxo-4-aza-5alpha-androst-1-en-17beta-carboxylic acid methyl ester, Methyl 3-oxo-4-aza-5alpha-androst-1-ene-17beta-carboxylate, Finasteride Impurity B
InChI Formula InChI=1S/C20H29NO3/c1-19-10-8-14-12(13(19)5-6-15(19)18(23)24-3)4-7-16-20(14,2)11-9-17(22)21-16/h9,11-16H,4-8,10H2,1-3H3,(H,21,22)/t12-,13-,14-,15+,16+,19-,20+/m0/s1
SMILES COC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name methyl (1S,3aS,3bS,5aR,9aR,9bS,11aS)-9a,11a-dimethyl-7-oxo-1,2,3,3a,3b,4,5,5a,6,9b,10,11-dodecahydroindeno[5,4-f]quinoline-1-carboxylate
Molecular Weight (g/mol) 331.45
Quantity 100 mg
Formula Weight 331.2147 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.