missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(furfurylthio)propionate, 98%, Thermo Scientific™

Product Code. 11340216
Click to view available options
Quantity:
1 g
25 g
100 g
Unit Size:
100g
1g
25g
This item is not returnable. View return policy

Product Code. 11340216

Brand: Thermo Scientific Alfa Aesar H32410.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94278-26-9
Molecular Formula C9H12O3S
Molecular Weight (g/mol) 200.252
MDL Number MFCD00053109
InChI Key MXXNUXUTQGTBGJ-UHFFFAOYSA-N
Synonym methyl 3-furfurylthio propionate, unii-l1387h8y3u, methyl furfuryl mercaptopropionate, methyl 3-2-furylmethylsulfanyl propanoate, 3-furfurylthio propanoic acid methyl ester, 3-furfurylthio propionic acid methyl ester, propanoic acid, 3-2-furanylmethyl thio-, methyl ester, methyl 3-furan-2-ylmethyl sulfanyl propanoate, methyl3-furfurylthio propionate
PubChem CID 15225386
IUPAC Name methyl 3-(furan-2-ylmethylsulfanyl)propanoate
SMILES COC(=O)CCSCC1=CC=CO1

Specifications

Density 1.163
Boiling Point 114°C to 116°C (1.12 mmHg)
Flash Point 92°C (198°F)
Refractive Index 1.512
Quantity 1 g
Formula Weight 200.26
Percent Purity 98%
Chemical Name or Material Methyl 3-(furfurylthio)propionate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.