accessibility menu, dialog, popup

UserName

CAS RN 94278-26-9

H3COOSO

CAS RN 94278-26-9

IUPAC Name: methyl 3-{[(furan-2-yl)methyl]sulfanyl}propanoate
Synonyms: methyl 3-[(furan-2-ylmethyl)sulfanyl]propanoate
Molecular Weight (g/mol): 200.25
Molecular Formula: C9H12O3S
InChi Key: MXXNUXUTQGTBGJ-UHFFFAOYSA-N
SMILES: COC(=O)CCSCC1=CC=CO1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)