missing translation for 'onlineSavingsMsg'
Learn More

Methyl 3-(4-methyl-1-piperazinyl)benzoate, 97%, Thermo Scientific™

Product Code. 15432118
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432118 5 g 5g
15422118 1 g 1g
15442118 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15432118 Supplier Thermo Scientific Alfa Aesar Supplier No. H55770.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 474334-89-9
Molecular Formula C13H18N2O2
Molecular Weight (g/mol) 234.30
MDL Number MFCD07369792
InChI Key LPOJJZXQESTVEV-UHFFFAOYSA-N
Synonym methyl 3-4-methylpiperazin-1-yl benzoate, methyl 3-4-methyl-1-piperazinyl benzoate, 1-3-carbomethoxyphenyl-4-methylpiperazine, 1-3-carbomethoxyphenyl-4-methylpipera, methyl 3-4-methylpiperazino benzoate
PubChem CID 16217966
IUPAC Name methyl 3-(4-methylpiperazin-1-yl)benzoate
SMILES COC(=O)C1=CC(=CC=C1)N1CCN(C)CC1

Specifications

Color Colorless to Yellow
Density 1.119 g/mL
Boiling Point 169°C to 172°C (0.9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.563
Quantity 5 g
Formula Weight 234.3
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl 3-(4-methyl-1-piperazinyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.