accessibility menu, dialog, popup

UserName

CH3NNOOH3C

CAS RN 474334-89-9

IUPAC Name: methyl 3-(4-methylpiperazin-1-yl)benzoate
Synonyms: methyl 3-(4-methylpiperazin-1-yl)benzoate
Molecular Weight (g/mol): 234.3
Molecular Formula: C13H18N2O2
InChi Key: LPOJJZXQESTVEV-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=CC=C1)N1CCN(C)CC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results