missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-(trifluoromethyl)benzoate, 97%, Thermo Scientific™

Product Code. 11398985
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398985 1 g 1g
11308995 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398985 Supplier Thermo Scientific Alfa Aesar Supplier No. L14132.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-96-7
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.148
MDL Number MFCD00236050
InChI Key JLCAMDSWNZBTEE-UHFFFAOYSA-N
Synonym methyl 2-trifluoromethyl benzoate, benzoic acid, trifluoromethyl-, methyl ester, acmc-1ahfp, methyl o-trifluoromethylbenzoate, jlcamdswnzbtee-uhfffaoysa, methyl-2-trifluoromethyl benzoate, 2-trifluoromethylbenzoic acid methyl ester, 2-trifluoromethyl-benzoic acid methyl ester, 2-trifluoromethyl benzoic acid methyl ester, methyl alpha,alpha,alpha-trifluoro-o-toluate
PubChem CID 2775578
IUPAC Name methyl 2-(trifluoromethyl)benzoate
SMILES COC(=O)C1=CC=CC=C1C(F)(F)F

Specifications

Density 1.306
Boiling Point 104°C to 105°C (25 mmHg)
Refractive Index 1.455
Quantity 1 g
Beilstein 2580873
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material Methyl 2-(trifluoromethyl)benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.