Learn More
Methyl 2-chloro-6-methoxypyridine-4-carboxylate, 98%
Product Code. 15468465
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468465 25 g 25g
15456247 1 g 1g
15458465 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15468465 Supplier Thermo Scientific Alfa Aesar Supplier No. H33438.14
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
CAS: 42521-10-8 | C8H8ClNO3 | 201.606 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 42521-10-8
Molecular Formula C8H8ClNO3
Molecular Weight (g/mol) 201.606
MDL Number MFCD00173929
InChI Key DAPAXVAUEVRBGS-UHFFFAOYSA-N
Synonym methyl 2-chloro-6-methoxyisonicotinate, 2-chloro-6-methoxy-isonicotinicacidmethylester, 2-chloro-6-methoxy-4-methoxycarbonyl pyridine, methylchloromethoxyisonicotinate, methyl-2-chloro-6-methoxyisonicotinate, 2-chloro-6-methoxy-isonicotinic acid methyl ester, methyl 2-chloro-6-methoxypyridin-4-yl carboxylate, 2-chloro-6-methoxy-isonicotinic acidmethyl ester, 4-pyridinecarboxylic acid, 2-chloro-6-methoxy-, methyl ester
PubChem CID 13339618
IUPAC Name methyl 2-chloro-6-methoxypyridine-4-carboxylate
SMILES COC1=NC(=CC(=C1)C(=O)OC)Cl
Specifications
Melting Point 103°C to 105°C
Quantity 25 g
Formula Weight 201.61
Percent Purity 98%
Chemical Name or Material Methyl 2-chloro-6-methoxypyridine-4-carboxylate