accessibility menu, dialog, popup

UserName

CH3ONOOH3CCl

CAS RN 42521-10-8

IUPAC Name: methyl 2-chloro-6-methoxypyridine-4-carboxylate
Synonyms: methyl 2-chloro-6-methoxypyridine-4-carboxylate
Molecular Weight (g/mol): 201.61
Molecular Formula: C8H8ClNO3
InChi Key: DAPAXVAUEVRBGS-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OC)=NC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results