Learn More
Methyl 2-(4-Butoxyphenyl)acetate, Mikromol™
Product Code. 30052292
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30052292 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052292 Supplier Mikromol Supplier No. MM0179.02
 Restricted Item
Estimated Shipment: 30-10-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Methyl 2-(4-Butoxyphenyl)acetate
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methyl 2-(4-Butoxyphenyl)acetate
CAS 29056-06-2
Analyte or Component Names Methyl 2-(4-Butoxyphenyl)acetate
Shipping Condition Room Temperature
Molecular Formula C13 H18 O3
Certifications/Compliance ISO 17025
Synonym Methyl 2-(4-Butoxyphenyl)acetate, Bufexamac Imp. B (EP), Bufexamac Impurity B
InChI Formula InChI=1S/C13H18O3/c1-3-4-9-16-12-7-5-11(6-8-12)10-13(14)15-2/h5-8H,3-4,9-10H2,1-2H3
SMILES CCCCOc1ccc(CC(=O)OC)cc1
IUPAC Name methyl 2-(4-butoxyphenyl)acetate
Molecular Weight (g/mol) 222.28
Quantity 100 mg
Formula Weight 222.1256 g/mol