Learn More
Methyl 1-naphthylacetate, 97%, Thermo Scientific™
Product Code. 11434484
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434484 100 g 100g
11414484 5 g 5g
11424484 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11434484 Supplier Thermo Scientific Alfa Aesar Supplier No. A13126.22
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Methyl 1-naphthylacetate, is also used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Sparingly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2876-78-0
Molecular Formula C13H12O2
Molecular Weight (g/mol) 200.237
MDL Number MFCD00004045
InChI Key YGGXZTQSGNFKPJ-UHFFFAOYSA-N
Synonym methyl 1-naphthaleneacetate, methyl 1-naphthylacetate, hormonit, kartofin, methyl 2-naphthalen-1-yl acetate, 1-naphthaleneacetic acid methyl ester, m 1 growth regulator, 1-naphthaleneacetic acid, methyl ester, methyl alpha-naphthylacetate, methyl naphthaleneacetic acid
PubChem CID 17891
IUPAC Name methyl 2-naphthalen-1-ylacetate
SMILES COC(=O)CC1=CC=CC2=CC=CC=C21
Specifications
Density 1.144
Boiling Point 155°C to 157°C (9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.598
Quantity 100 g
Beilstein 1956293
Solubility Information Sparingly soluble in water.
Formula Weight 200.24
Percent Purity 97%
Chemical Name or Material Methyl 1-naphthylacetate