Learn More
SPEX CertiPrep™ Method EN15662 SPEXQuE™ QuEChERS Kit
Product Code. 15286036 Shop All SPEX Certiprep Products
Click to view available options
Volume (Metric):
2 mL
50 mL
Quantity:
100 Pack
50 Pack
Unit Size:
1 unit
Pack of 100
Pack of 50
Product Code. 15286036
Brand: SPEX CertiPrep™ ENPSA2ML
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Meet your QuEChERS needs with these general purpose kits
Meeting all your QuEChERS needs with general purpose kits suitable for removal of polar organic acids, some low to moderate levels of sugars and lipids.
  • European Methods (EN Method 15662)
Specifications
Sorbent 25 mg PSA, 150 mg MgSO4
Type PSA Dispersive SPE Clean-up Tube
Volume (Metric) 2 mL
Method EN 15662
Quantity 100 Pack