missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methicillin-resistant Staphylococcus aureus Mouse anti-Staphylococcus, Clone: 332/423, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15091.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against Methicillin-resistant Staphylococcus aureus.
- High specificity
- High reactivity
Specifications
| Methicillin-resistant Staphylococcus aureus | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1-5 ug/mL for coating) Immunofluorescence (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| Staphylococcus species | |
| Liquid |
| ELISA, Immunofluorescence | |
| 332/423 | |
| Mouse monoclonal antibody raised against Methicillin-resistant Staphylococcus aureus. | |
| In 10 mM PBS. | |
| Methicillin-resistant Staphylococcus aureus. | |
| 100 μg | |
| Primary | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction