Learn More
Methenolone enantate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Primobolan |
| CAS | 303-42-4 |
| Analyte or Component Names | Primobolan |
| Shipping Condition | Room Temperature |
| Molecular Formula | C27 H42 O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Metenolone enanthate, Methenolone 17-enanthate, Methenolone enanthate, NSC 64967, Primobolan Depot, Primonabol Depot, SH 601, SQ 16374, (5alpha,17beta)-1-Methyl-17-[(1-oxoheptyl)oxy]-androst-1-en-3-one, 5alpha-Androst-1-en-3-one, 17beta-hydroxy-1-methyl-, heptanoate (6CI,8CI), (5alpha,17beta)-1-Methyl-17-[(1-oxoheptyl)oxy]androst-1-en-3-one, 1-Methyl-17beta-hydroxy-5alpha-androst-1-en-3-one enanthate, 1-Methyl-Delta1-androsten-17beta-ol-3-one enanthate, 17beta-Hydroxy-1-methyl-5alpha-androst-1-en-3-one 17beta-enanthate, 17beta-Hydroxy-1-methyl-5alpha-androst-1-en-3-one heptanoate |
| InChI Formula | InChI=1S/C27H42O3/c1-5-6-7-8-9-25(29)30-24-13-12-22-21-11-10-19-17-20(28)16-18(2)27(19,4)23(21)14-15-26(22,24)3/h16,19,21-24H,5-15,17H2,1-4H3/t19-,21-,22-,23-,24-,26-,27-/m0/s1 |
| SMILES | CCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)C=C(C)[C@]4(C)[C@H]3CC[C@]12C |
| Show More |