missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Metapneumovirus Glycoprotein G Antibody [HRP] - (HMPV/Homo sapiens/PER/CFI0320/2010/A), Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43773H
This item is not returnable.
View return policy
Description
Metapneumovirus Glycoprotein G Polyclonal antibody specifically detects Metapneumovirus Glycoprotein G in Virus samples. It is validated for ELISA,Western Blot
Specifications
| Metapneumovirus Glycoprotein G | |
| Polyclonal | |
| Western Blot, ELISA | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| HRP | |
| PBS | |
| Produced in rabbits immunized with purified, recombinant Human Metapneumovirus Glycoprotein G (HMPV/Homo sapiens/PER/CFI0320/2010/A) (RefSeq#: AHV79442.1; Asn52-Asn228) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction