missing translation for 'onlineSavingsMsg'
Learn More

Metapneumovirus Antibody (hMPV57) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

214.00€ - 440.00€

Specifications

Antigen Metapneumovirus
Clone hMPV57
Concentration 1.0 mg/mL
Dilution Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18240581
View Documents
Novus Biologicals
NBP2-53105
0.1 mL
440.00€
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18685907
View Documents
Novus Biologicals
NBP2-53105-0.025ml
0.025 mL
214.00€
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Metapneumovirus Monoclonal specifically detects Metapneumovirus in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Metapneumovirus
1.0 mg/mL
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Unconjugated
Mouse
Virology Bacteria and Parasites
PBS with 0.02% Sodium Azide
IgG1
Protein A purified
hMPV57
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500
Monoclonal
Purified
RUO
Virus
Human MPV strain NCL-174
Primary
Identifies Metapneumovirus nucleoprotein of both types A and B.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.