Learn More
Toxaphene in DCM 100μg/mL, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Packaging | Ampoule |
| CAS | 8001-35-2 |
| Concentration | 100 μg/mL |
| Type | Pesticide and Metabolite Single-component Standard Solution |
| Solvent or Matrix | Dichloromethane |
| Quantity | 1 mL |
| Chemical Name or Material | Toxaphene |