Learn More
Metabolite Dibenzo(a,h)anthracene in DCM 100μg/mL, Fisher Chemical™
Product Code. 12945294 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12945294 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12945294 Supplier Fisher Chemical Supplier No. P816960
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule(s)
CAS 53-70-3
Molecular Formula C22H14
Concentration 100 μg/mL
InChI Key LHRCREOYAASXPZ-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)C=CC3=CC4=C(C=CC5=CC=CC=C54)C=C32
IUPAC Name naphtho[1,2-b]phenanthrene
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 278.354
PubChem CID 5889
Quantity 1 mL
ChEBI CHEBI:35299
Chemical Name or Material Dibenzo(a,h)anthracene