Learn More
Bis-(4-aminophenyl)-methane in Toluene 100μg/mL, Fisher Chemical™
Product Code. 12963184 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12963184 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12963184 Supplier Fisher Chemical Supplier No. P874420
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Packaging Ampoule
CAS 101-77-9
Molecular Formula C13H14N2
Concentration 100 μg/mL
MDL Number MFCD00007914
InChI Key YBRVSVVVWCFQMG-UHFFFAOYSA-N
SMILES NC1=CC=C(CC2=CC=C(N)C=C2)C=C1
IUPAC Name 4-[(4-aminophenyl)methyl]aniline
Solvent or Matrix Toluene
Molecular Weight (g/mol) 198.27
PubChem CID 7577
Quantity 1 mL
ChEBI CHEBI:32506
Chemical Name or Material Bis-(4-aminophenyl)-methane