Learn More
Bioresmethrin in Methanol 100μg/mL, Fisher Chemical™
Product Code. 12986963 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12986963 10 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12986963 Supplier Fisher Chemical Supplier No. P879300
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Packaging Ampoule
CAS 28434-01-7
Molecular Formula C22H26O3
Concentration 10 μg/mL
InChI Key VEMKTZHHVJILDY-UXHICEINSA-N
Type Pesticide and Metabolite Standards
SMILES CC(=CC1C(C1(C)C)C(=O)OCC2=COC(=C2)CC3=CC=CC=C3)C
IUPAC Name (5-benzylfuran-3-yl)methyl (1R,3R)-2,2-dimethyl-3-(2-methylprop-1-enyl)cyclopropane-1-carboxylate
Solvent or Matrix Iso-octane
Molecular Weight (g/mol) 338.447
PubChem CID 162381
Quantity 10 mL
ChEBI CHEBI:39280
Chemical Name or Material Bioresmethrin