Learn More
Metabolite 2-Fluorophenol in DCM 100μg/mL, Fisher Chemical™
Product Code. 12946783 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12946783 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12946783 Supplier Fisher Chemical Supplier No. P826520
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging Ampoule(s)
CAS 367-12-4
Molecular Formula C6H5FO
Concentration 100 μg/mL
MDL Number MFCD00002155
InChI Key HFHFGHLXUCOHLN-UHFFFAOYSA-N
SMILES OC1=CC=CC=C1F
IUPAC Name 2-fluorophenol
Solvent or Matrix Dichloromethane
Molecular Weight (g/mol) 112.10
PubChem CID 9707
Quantity 1 mL
Chemical Name or Material 2-Fluorophenol